Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCGB1D2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548633

Catalog No. PA548633

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with rat based on sequence homology.

May bind androgens and other steroids, may also bind estramustine, a chemotherapeutic agent used for prostate cancer. May be under transcriptional regulation of steroid hormones.
Specifications

Specifications

SCGB1D2
Polyclonal
PBS with 0.09% sodium azide
O95969
Secretoglobin family 1D member 2, Lipophilin-B, SCGB1D2, LIPHB, LPNB
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10647
Western Blot
Unconjugated
SCGB1D2
Liquid
SCGB1D2
KLH conjugated synthetic peptide between 22-50 amino acids from the Central region of human SCGB1D2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit