Learn More
SCGF beta Mouse anti-Human, Alexa Fluor 488, Clone: MM0533-6D5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211874AF488
Catalog No. NB211874488
Add to cart
Description
Description
SCGF/CLEC11a Monoclonal specifically detects SCGF/CLEC11a in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
SCGF beta
MM0533-6D5
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
CLEC11A
Human SCGF beta
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
CLECSF3C-type lectin superfamily member 3, C-type lectin domain family 11 member A, C-type lectin domain family 11, member A, LSLCLC-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 3, Lymphocyte secreted C-type lectin, lymphocyte secreted long form of C-type lectin, p47, SCGF, Stem cell growth factor, stem cell growth factor; lymphocyte secreted C-type lectin
Mouse
IgG1
0.1 ml
Primary
6320