Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCH 529074, Tocris Bioscience™

Catalog Number 424010
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
922150-11-6
C31H36Cl2N6
563.571
NCAJLQDPTZBGJV-UHFFFAOYSA-N
n'-2-4-bis-4-chlorophenyl methyl piperazin-1-ylmethyl quinazolin-4-yl-n,n-dimethylpropane-1,3-diamine, n3-2-4-bis 4-chlorophenyl methyl-1-piperazinyl methyl-4-quinazolinyl-n1,n1-dimethyl-1,3-propanediamine
12001922
N-[2-[[4-[bis(4-chlorophenyl)methyl]piperazin-1-yl]methyl]quinazolin-4-yl]-N',N'-dimethylpropane-1,3-diamine
CN(C)CCCNC1=NC(=NC2=CC=CC=C21)CN3CCN(CC3)C(C4=CC=C(C=C4)Cl)C5=CC=C(C=C5)Cl
This item is not returnable. View return policy
922150-11-6
C31H36Cl2N6
563.571
NCAJLQDPTZBGJV-UHFFFAOYSA-N
n'-2-4-bis-4-chlorophenyl methyl piperazin-1-ylmethyl quinazolin-4-yl-n,n-dimethylpropane-1,3-diamine, n3-2-4-bis 4-chlorophenyl methyl-1-piperazinyl methyl-4-quinazolinyl-n1,n1-dimethyl-1,3-propanediamine
12001922
N-[2-[[4-[bis(4-chlorophenyl)methyl]piperazin-1-yl]methyl]quinazolin-4-yl]-N',N'-dimethylpropane-1,3-diamine
CN(C)CCCNC1=NC(=NC2=CC=CC=C21)CN3CCN(CC3)C(C4=CC=C(C=C4)Cl)C5=CC=C(C=C5)Cl

Restores mutant p53 activity

Restores mutant p53 activity. Binds p53 DNA binding domain (Kd = 1-2 μM) and restores wild-type function to many oncogenic mutants by acting as a chaperone. Inhibits ubiquitination of p53 by HDM2. Triggers apoptosis in tumor cell lines and reduces tumor growth in a xenograft model.
Quantity 10 mg
Formula Weight 563.56
Percent Purity >98%
Chemical Name or Material SCH 529074
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit