Learn More
Azzota Corp SCH-772984 DFS Item
Manufacturer:  Azzota Corp C1298
SCH-772984 is a potent and selective ERK inhibitor with potential anticancer activity. SCH-722984 showed activity against BRAF mutant, NRAS mutant and wild-type melanoma. Please request if need more quantities
Catalog No. 50-161-3182