Learn More
SCHIP1 Mouse anti-Canine, Human, Mouse, Non-human primate, Rat, Clone: OTI1C4, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525917
Catalog No. PIMA525917
Add to cart
Description
Description
This locus represents naturally occurring read-through transcription from the neighboring IQ motif containing J (IQCJ) and schwannomin interacting protein 1 (SCHIP1) genes. Alternative splicing results in multiple transcript variants that are composed of in-frame exons from each individual gene. The resulting fusion products are thought to be components of the multimolecular complexes of axon initial segments and nodes of Ranvier, and they may play a role in calcium-mediated responses.
Specifications
Specifications
SCHIP1
Monoclonal
0.4 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q3TI53, Q9P0W5
Schwannomin-interacting protein 1, SCHIP-1, SCHIP1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
295105, 29970, 30953, 478678, 703321
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1C4
Unconjugated
SCHIP1
Liquid
LOC478678, Schip1
Full length human recombit protein of SCHIP1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Non-human primate, Rat