Learn More
Science First™ Center of Gravity Paradox
Things are not always what they seem.
Supplier: Science First™ 6110370
Description
- The angular momentum apparatus is weighted at one end
- One meter rod is tried to balance with heavier end down
- Intuition tells that most people will think it should work
- In actuality, opposite is true
- The rod is turnedover and balanced on a mere fingertip
- A hands-on way to introduce the concepts of the center of gravity
- Angular acceleration and moment of inertia
- Movable mass, instructions include theory
Specifications
Center of Gravity Paradox | |
For weighing angular momentum apparatus |