Learn More
Science First™ Heat Absorption Comparator Products for Science Education
Compare heat absorption and radiation.
Manufacturer:  Science First™ 6120050
Catalog No. S98908
Add to cart
Includes: Two pans (90mm diameter), one dark, one silver
Description
Description
  • One 10W/12V lamp mounted between the pans shows different temperature increases
  • With binding posts and instructions on the base
  • Requires two thermometers
Specifications
Specifications
Two pans, one dark, one silver
Heat absorption comparator