Scientific Cell Single Filament Hollow Cathode Lamps
Manufacturer: Scientific Cell 5BA/KA
 View more versions of this product
Catalog No. 14-386-149
  Check Availability
Add to Cart
    Specifications
    Diameter (English) Outer 2 in.
    Diameter (Metric) Outer 50mm
    Filament Potassium
    For Use With (Equipment) Perkin-Elmer™ instruments
    Light Source Hollow Cathode
    Product Type Single Filament Hollow Cathode Lamp
  • Description & Specifications
Related Products
Product Content Feedback