Learn More
Scientific Device Closed System Fecal Concentrators
Allows more safe method with minimum of handling and odor
Manufacturer:  Scientific Device Laboratory, IncSupplier Diversity Partner 6056
 View more versions of this product
Catalog No. 23-769-517
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Preassembled tube-strainer unit, fecal Mucolytic reagent, filter with 0.6mm holes, dispensing spoons, processing trays and applicator sticks
Fecal Parasite Concentrator kit
120 Pack