Learn More
Scientific Device Closed System Fecal Concentrators
Allows more safe method with minimum of handling and odor
$402.00
Specifications
Includes Preassembled tube-strainer unit, fecal Mucolytic reagent, filter with 0.6mm holes, dispensing spoons, processing trays and applicator sticks
Product Type Fecal Parasite Concentrator kit
View More Specs
Includes: Pre-assembled tube-strainer unit, fecal mucolytic reagent, dispensing spoons (for 15mL tubes), processing trays, and applicator sticks
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-769-516
View Documents
Scientific Device
6052
100 Pack
Pack of 100 for $402.00
Add to cart
23-769-517
View Documents
Scientific Device
6056
120 Pack This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Filter posses 0.6mm holes that allows parasites to pass through but block larger fecal material
  • Available in tube size of 15mL and 50mL
  • For use with safe, formalin-free fixative parasafe
Specifications
Specifications
Preassembled tube-strainer unit, fecal Mucolytic reagent, filter with 0.6mm holes, dispensing spoons, processing trays and applicator sticks
Fecal Parasite Concentrator kit