Scientific Device Cryptosporidium Control Slides
Many typical organisms on each slide
Manufacturer: Scientific Device Laboratory, IncSupplier Diversity Partner 37050
 View more versions of this product
Catalog No. 23-769-302
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Quantity 50 Pack
    Type Cryptosporidium Control Slide
    Includes Positive and negative well on each slide
    For Use With (Application) Used for staining both cryptosporidium and nocardia
    • Cryptosporidium sps. is now recognized as an important intestinal pathogen in patients with HIV infection
    • Routine parasitological techniques fail to demonstrate this protozoa
    • Modified Acid Fast staining techniques prove effective in staining this organism
    • Each package contains 10 slides, each having a positive and negative well
  • Description & Specifications
Related Products
Product Content Feedback