Scientific Device McFarland Standards
Available in McFarland or Colorimeter Standard Sets
Manufacturer: Scientific Device Laboratory, IncSupplier Diversity Partner 2370
 View more versions of this product
Catalog No. 23-769-316
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Includes 0.5, 1, 2 and 3 Standard
    Product Type Colorimeter standard set
    Quantity 4 Pack
    Shelf Life 2 Years
    Storage Requirements Room temperature
    • Each particle is the size of Entamoeba Coli
    • Each tube contains 12mL (.40 oz.) of suspension
    • Latex particle suspension
    • Extended shelf life
  • Description & Specifications
Related Products
Product Content Feedback