Scientific Device Protozoa Stain Control Slides
Ideal for control of Trichrome or Hematoxylin staining procedures
Manufacturer: Scientific Device Laboratory, IncSupplier Diversity Partner 354ZN
 View more versions of this product
Catalog No. 23-769-526
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    For Use With (Application) For trichrome or hematoxylin staining procedures
    Includes Smear of clinical protozoans in PVA, fixed with parasafe, copper/zinc
    Quantity 10 Pack
    Type Protozoa Stain Control Slide with Zinc
  • Description & Specifications
Related Products
Product Content Feedback