VIP-Fixative™ Forma-Vial™ Pre-filled 10% Neutral Buffered Formalin Specimen Containers

Safely transport, store, and retrieve pathology specimens. Scigen™ VIP-Fixative™ Forma-Vial™ Pre-filled 10% Neutral Buffered Formalin Specimen Containers are available in a variety of sizes.

Manufacturer: SciGen IncSupplier Diversity Partner 1007

 View more versions of this product

Catalog No. 23-730-585

  Check Availability
Add to Cart

    Specifications

    For Use With Histology/cytology
    Graduated No
    Capacity 7mL, 0.2 oz.
    Volume (English) 0.2 oz.
    Color Clear
    Volume (Metric) 7mL
    No. per Case 400
    Closure Included Yes
    Product Type Specimen Container, Vial
    Material Polypropylene

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit