Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Scigen VIP-Fixative Leak-Proof Specimen Containers Buffered Zinc 10% Formalin Filled
Safely transport, store and retrieve pathology specimens using Scigen's Forma-Vial™ system. Containers from 30 mL to 7.5 Liters, filled with buffered zinc formalin, OSHA compliant.
Supplier: Scigen 6500Z
Description
- Forma-Vial™ specimen containers are uniquely designed to be leak proof and protect against biohazardous or hazardous material release to the environment Highly visible OSHA compliant labeling
- Sizes range from 30mL specimen containers to 7.5 Liters operating room organ buckets
- Container openings range from 50 mm to 300 mm
- Filled with buffered zinc formalin to 50% of container’s capacity
- Buffered zinc 10% formalin to enhance nuclear detail in immunohistochemistry and special staining techniques
- Chloride, sulfate and mercury free
- Nonsterile containers marked with a lot code and expiration date, 4 year shelf life
- Formalin waste designed to be treated in bulk using Neutralex or treated individually using Neutralex tablets (patent pending).
- Manufactured using ISO 13485 and FDA quality systems
Specifications
Specimen Container Bucket | |
No | |
Non-sterile | |
7500 mL | |
HDPE | |
Valve Cap | |
Specimen Container Pre-filled With Zinc Buffered 10% Formalin | |
Histology/cytology specimen containers | |
267 mm | |
OSHA Compliant | |
1 Ea. |
Yes | |
HDPE | |
2 gal. | |
White | |
292 mm | |
White | |
254 mm | |
10.5 in. | |
50% Filled Zinc Buffered 10% Formalin | |
1 Ea. | |
Forma-Vial™ Specimen Containers |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction