Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scigen Neutralex: Grossing Pads Neutra-Pads

Grossing pads instantaneously neutralize formalin, while rapidly absorbing harmful liquids. Blue cut resistance surface with plastic backing keeps your grossing area or storage area clean and dry.

$88.10 - $451.50

Specifications

Capacity Neutralizes and Absorbs 0.05 gal., 180mL 10% Formalin
Product Type Neutra-Pads™
For Use With (Application) For Formalin, Grossing and Storage Safety
View More Specs

Products 3
Catalog Number Mfr. No. Dimensions (L x W) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Quantity Price Quantity & Availability  
23-730-568
View Documents
Scigen
42951
8 x 10 in. 10 Pads/cs. Pack of 10 for $88.10
Add to cart
 
23-730-570
View Documents
Scigen
4297
11 x 17 in. 15 Pads/Cs. Case of 15 for $94.10
Add to cart
 
23-730-567
View Documents
Scigen
4295
8 x 10 in. 100 Pads/cs. Pack of 100 for $451.50
Add to cart
 
Description

Description

  • Neutralizes formalin on-contact while absorbing hazardous liquids
  • Highly absorbent polymeric fiber embedded with formalin neutralizer
  • Eliminates hazardous vapors
  • Blue dissection surface consists of a porous highly-resilient polyethylene copolymer cutting area
  • Plastic backing prevents the escape of harmful liquids, keeping you grossing or storage area clean and dry
  • Available in two sizes: 8” x 10” or 11” x 17”
  • Each 8” x 10”pad absorbs up to 80mL of formalin
  • Each 11” x 17” pad absorbs up to 150mL of formalin
  • Manufactured using ISO 13485 quality system

Specifications

Specifications

Neutralizes and Absorbs 0.05 gal., 180mL 10% Formalin
Neutra-Pads™
For Formalin, Grossing and Storage Safety
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit