Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scigen Neutralex Certified Formalin Neutralizer

Treat Formalin waste onsite with this Ca EPA validated pollution prevention technology. Easy-to-use, pre-measured powder treats formalin in 15 minutes

Manufacturer:  Scigen 4047

 View more versions of this product

Catalog No. 23-730-561


Add to cart

Description

Description

  • Easy-to-use, pre-measured powder treats formalin in 5 to 15 minutes with no drain clogging polymers, no obnoxious vapors, and no pH adjustment
  • Neutralex has been certified for down-the drain disposal for more than 20 years
  • Product and process for onsite formalin treatment is validated by California Environmental Protection Agency as a Pollution Prevention Technology
  • Authorized under Health and Safety Code 25200.1.5, 25200.3.1 and 22CCR 67450.20 for onsite treatment of formalin waste
  • Neutralex treated waste does not pose a hazard to public health and safety or to the environment
  • Treated formalin waste has a residual formaldehyde concentration of less than 1 part per million and pH between 6 and 9
  • Eliminates the need for Hazardous waste hauling
  • More than one million gallons of formalin waste treated with 100% compliance
  • Manufactured using ISO 13485 quality system

Specifications

Specifications

CalEPA Certified Polution Prevention Technology
For neutralization of formalin waste, glutaraldehyde, paraldehyde and OPA solutions
16 Pack
12.7kg
Healthcare
Neutralizes 4L of 10% Formalin Waste
Pre-measured pouches
28lbs
Certified Formalin Neutralizer
Safety and Handling

Safety and Handling

ShelfLife : 4 Years

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit