Learn More
Scigen Neutralex™ Certified Formalin Neutralizer
Environmentally safe and certified technology for neutralization of formalin waste
Manufacturer:  SciGen IncSupplier Diversity Partner 4048
 View more versions of this product
Catalog No. 23-730-562
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
55.11 lb.
55.11 lb., 25kg
For neutralization of formalin waste
Each
Certified Formalin Neutralizer
25kg
Neutralizes 150L
25kg pail
25kg