Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SciMatCo™ Nonmetallic Flammables Cabinets
SCI_ED

Protect contents as well as metal cabinets.

Manufacturer:  SciMatCo™ SC7131/EMD

 View more versions of this product

Catalog No. S94940MF


Add to cart

Includes: Lock and two keys

Description

Description

Wooden flammables cabinets constructed from 1 in. thick (2.5cm) 9-ply plywood, using specifications developed by both OSHA and NFPA.

  • Unlike metal, wood provides good thermal insulation and won't bend or distort when exposed to heat
  • All sizes have continuous piano-style hinges on doors and liquid-tight trough floors
  • All joints are rabbeted for strength, and cabinets are finished with yellow epoxy paint inside and out
  • Cabinets have 10 × 10 in. (25 × 25cm) “FLAMMABLE” hazard label on door(s), and a “KEEP FIRE AWAY” warning
  • Adjustable shelves feature metal shelf standards and supports
  • All hardware is corrosion resistant

Order Info

Four and 18-gallon cabinets have one door and one adjustable shelf; 45-gallon cabinet has two doors and two adjustable shelves.

Compliance

Meet all applicable OSHA 1910 and NFPA 30 standards.

Specifications

Specifications

15.12 x 41 x 42 in. (38 x 104.14 x 106.68 cm)
109 cm
104.14 cm
2
45 gal.
Manual-close
41 in.
Wood (Plywood)
44.6 in.
Flammables
15.12 in.
106.68 cm
38 cm
58.42 cm
113.28 cm
Storage Cabinet
23.8 in.
Yellow
OSHA, NFPA
43 in.
Internal Metal lock with two keys, piano type door hinge, 2 in. liquid tight trough
42 in.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit