Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCIMP Mouse anti-Human, Alexa Fluor™ 647, Clone: NVL-07, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 568955
This item is not returnable.
View return policy
Description
SCIMP Monoclonal antibody specifically detects SCIMP in Human samples. It is validated for Intracellular staining (flow cytometry) (Routinely Tested)Specifications
SCIMP | |
Monoclonal | |
Alexa Fluor 647 | |
SLP adaptor and CSK interacting membrane protein | |
Human Recombinant intracellular part of human SCIMP. | |
25 Tests | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
NVL-07 | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
388325 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title