Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ScintiVerse™ LC Cocktail (Scintanalyzed™), Fisher Chemical

Catalog No. SX174
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle

Special-purpose scintillation cocktail for measuring radio-tagged analytes in HPLC eluants containing organic solvents

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ScintiVerse™ LC Cocktail
Physical Form Liquid
CAS 95-63-6, 68131-39-5, 67-63-0, 92-71-7, 1806-34-4
Background 50 CPM max.
Color Blue
Quantity 4 L
MDL Number MFCD00212672
Packaging Glass Bottle
Grade Scintanalyzed
Specific Gravity 0.880 to 0.900 (at 25°C)
Water record%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.