Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCIO 469 hydrochloride, Tocris Bioscience™

Catalog Number p-4816849
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
309913-83-5
C27H30ClFN4O3
513.01
ZMELOYOKMZBMRB-DLBZAZTESA-N
talmapimod, scio-469, scios 469, scio 469, unii-b1e00kq6nt, b1e00kq6nt, scio 469 hydrochloride, 2-6-chloro-5-2r,5s-4-4-fluorobenzyl-2,5-dimethylpiperazin-1-yl carbonyl-1-methyl-1h-indol-3-yl-n,n-dimethyl-2-oxoacetamide, talmapimod usan, talmapimod usan:inn
9871074
2-[6-chloro-5-[(2R,5S)-4-[(4-fluorophenyl)methyl]-2,5-dimethylpiperazine-1-carbonyl]-1-methylindol-3-yl]-N,N-dimethyl-2-oxoacetamide
CC1CN(C(CN1C(=O)C2=C(C=C3C(=C2)C(=CN3C)C(=O)C(=O)N(C)C)Cl)C)CC4=CC=C(C=C4)F
This item is not returnable. View return policy
309913-83-5
C27H30ClFN4O3
513.01
ZMELOYOKMZBMRB-DLBZAZTESA-N
talmapimod, scio-469, scios 469, scio 469, unii-b1e00kq6nt, b1e00kq6nt, scio 469 hydrochloride, 2-6-chloro-5-2r,5s-4-4-fluorobenzyl-2,5-dimethylpiperazin-1-yl carbonyl-1-methyl-1h-indol-3-yl-n,n-dimethyl-2-oxoacetamide, talmapimod usan, talmapimod usan:inn
9871074
2-[6-chloro-5-[(2R,5S)-4-[(4-fluorophenyl)methyl]-2,5-dimethylpiperazine-1-carbonyl]-1-methylindol-3-yl]-N,N-dimethyl-2-oxoacetamide
CC1CN(C(CN1C(=O)C2=C(C=C3C(=C2)C(=CN3C)C(=O)C(=O)N(C)C)Cl)C)CC4=CC=C(C=C4)F

Selective p38 MAPK inhibitor

SCIO 469 hydrochloride is a selective, ATP-competitive p38 inhibitor (IC50 = 9 nM for p38α in vitro). Displays approximately 10-fold selectivity for p38α over p38β and 2000-fold selectivity for p38α over 20 other kinases. Reduces p38α phosphorylation in multiple myeloma cells in vitro and in vivo; activity results in decreased tumor burden and angiogenesis in murine models of multiple myeloma. Also enhances bortezomib -induced cytotoxicity against multiple myeloma cells.
Quantity 10 mg
Formula Weight 549.46
Percent Purity >98%
Chemical Name or Material SCIO 469 hydrochloride
Recommended Storage Desiccate at +4°C.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit