Learn More
Sciteck™ AdultaCheck™ Adulteration Test Strips
The first dry chemistry test strip to ever test for creatinine and glutaraldehyde in urine
Manufacturer:  SCITECK DIAGNOSTICS INC 30250
 View more versions of this product
Catalog No. 23-424-4510
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Creatinine, Aldehydes, Nitrite, Chromate, Peroxidase, pH (normal, abnormal), Halogen, Specific Gravity, Oxidant
For reagent impregnated test pads for specific analytes
50 Tests