Sciteck™ AdultaCheck™ Adulteration Test Strips
The first dry chemistry test strip to ever test for creatinine and glutaraldehyde in urine
Manufacturer: SCITECK DIAGNOSTICS INC 310
 View more versions of this product
Catalog No. 23-424-4511
  Check Availability
Add to Cart
    Specifications
    Detectable Analytes Creatinine, Oxidants, Glutaraldehyde, pH
    For Use With (Application) For reagent impregnated test pads for specific analytes
    Quantity 2 x 15mL
  • Description & Specifications
Related Products
Product Content Feedback