Sciteck™ AdultaCheck™ Adulteration Test Strips
The first dry chemistry test strip to ever test for creatinine and glutaraldehyde in urine
Manufacturer: SCITECK DIAGNOSTICS INC 330
 View more versions of this product
Catalog No. 23-424-4513
  Check Availability
Add to Cart
    Specifications
    Detectable Analytes Creatinine, Aldehydes, Nitrite, Chromate, Peroxidase, pH (normal, abnormal), Halogen, Specific Gravity, Oxidant
    For Use With (Application) For reagent impregnated test pads for specific analytes
    Quantity 4 x 15mL
  • Description & Specifications
Related Products
Product Content Feedback