Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sciteck AUTOUA Liquid Urinalysis Reagents: Bilirubin/Urobilinogen Combination Calibrators
User-friendly, automated open channel anlayzer reagent system
$181.00 - $534.00
Specifications
Format | Liquid |
---|---|
Intended Use | Pharmaceuticals |
For Use With (Equipment) | Hitachi 717, 911, 747, olympus series, beckman and auto analyzer |
Storage Requirements | 2-10°C |
Catalog Number | Mfr. No. | Concentration | Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Type | Quantity | Price | Quantity & Availability | |||
23-424-454
|
Sciteck
212/21302 |
Bilirubin 1mg/dL, Urobilinogen 1mg/dL | Bilirubin, Urobilinogen Calibrators | 3 x 3mL |
Each for $181.00
|
|
23-424-453
|
Sciteck
212/2132S |
Bilirubin 1, 30mg/dL, Urobilinogen 1, 30mg/dL | Bilirubin, Urobilinogen Calibrators | 2 x (3 x 3)mL |
Pack of 2 for $534.00
|
|
Description
- For quantitative analysis of 12 analytes in human urine: pH, specific gravity, ketone bodies, blood (Hb), leukocyte esterase (LE), nitrite, protein, glucose, bilirubin, urobilinogen, beta-hydroxybutyrate and creatinine
- Compatible with Hitachi 717, 911, 747, Olympus series, Beckman and other auto-analyzers
- Use with Creatinine Calibrator 23-424-4534, and Zero Calibrator 23-424-4545
Specifications
Liquid | |
Pharmaceuticals | |
Hitachi 717, 911, 747, olympus series, beckman and auto analyzer | |
2-10°C |
U.S. Foreign Patents and Patents Pending: 5,955,374; 5,801,059; 5,801,060; 5,759,860; 5,776,780; 5,780,239; 5,516,700; 5,733,785; 5,736,408; 6,468,805; 5,753,451; 6,617,123; 6,537,823
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title