Learn More
Sciteck™ AUTOUA™ Liquid Urinalysis Reagents: Hemoglobin Calibrators
User-friendly, automated open channel anlayzer reagent system
Manufacturer:  SCITECK DIAGNOSTICS INC 2043S
 View more versions of this product
Catalog No. 23-424-4599
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Disclaimers:
U.S. Foreign Patents and Patents Pending: 5,955,374; 5,801,059; 5,801,060; 5,759,860; 5,776,780; 5,780,239; 5,516,700; 5,733,785; 5,736,408; 6,468,805; 5,753,451; 6,617,123; 6,537,823
Specifications Description & Specifications
Specifications
Hemoglobin calibrators
Hemoglobin (RBC) 100, 500 and 1000μg/dL
Hitachi 717, 911, 747, olympus series, beckman and auto analyzer
3 x 25mL
Liquid
Pharmaceuticals
2-10°C