Learn More
Sciteck™ AUTOUA™ Liquid Urinalysis Reagents: Hemoglobin Calibrators
User-friendly, automated open channel anlayzer reagent system
Manufacturer:  SCITECK DIAGNOSTICS INC 20405
 View more versions of this product
Catalog No. 23-424-4601
  Check Availability
Add to cart
Disclaimers:
U.S. Foreign Patents and Patents Pending: 5,955,374; 5,801,059; 5,801,060; 5,759,860; 5,776,780; 5,780,239; 5,516,700; 5,733,785; 5,736,408; 6,468,805; 5,753,451; 6,617,123; 6,537,823
Specifications Description & Specifications
Specifications
Hemoglobin calibrators
Hemoglobin (RBC) 500μg/dL
Hitachi 717, 911, 747, olympus series, beckman and auto analyzer
25mL
Liquid
Pharmaceuticals
2-10°C