Sciteck™ AUTOUA™ Liquid Urinalysis Reagents

User-friendly, automated open channel anlayzer reagent system

Manufacturer: SCITECK DIAGNOSTICS INC 20430

 View more versions of this product

Catalog No. 23-424-4583

  Check Availability
Add to Cart

Disclaimers:

U.S. Foreign Patents and Patents Pending: 5,955,374; 5,801,059; 5,801,060; 5,759,860; 5,776,780; 5,780,239; 5,516,700; 5,733,785; 5,736,408; 6,468,805; 5,753,451; 6,617,123; 6,537,823


    Specifications

    Product Type Hemoglobin (RBC) reagent
    Format Liquid
    For Use With (Application) Pharmaceuticals
    For Use With (Equipment) Hitachi 717, 911, 747, olympus series, beckman and auto analyzer
    Storage Requirements 2-10°C
    Quantity 600mL, 300mL,600mL, 300mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit