Sciteck™ SVT™ Adulteration Aldehydes Reagent

For use with automated systems for drugs of abuse adulteration testing

Manufacturer: SCITECK DIAGNOSTICS INC 123

 View more versions of this product

Catalog No. 23-424-4537

  Check Availability
Add to Cart

Disclaimers:

U.S. Patent No. 5,464,775. Other U.S. and Foreign Patents Pending


    Specifications

    Product Type Aldehyde low calibrator
    For Use With (Application) In-vitro diagnostic use to detect the presence of Aldehydes in solution, forensic/toxicology use
    For Use With (Equipment) Automated system for drugs of abuse adulteration testing
    Storage Requirements 2° to 10°C
    Quantity 60mL,60mL
    Source Human Urine

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit