Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sciteck SVT Adulteration Aldehydes Reagent
For use with automated systems for drugs of abuse adulteration testing
$344.00
Specifications
Source | Human Urine |
---|---|
Intended Use | In-vitro diagnostic use to detect the presence of Aldehydes in solution, forensic/toxicology use |
For Use With (Equipment) | Automated system for drugs of abuse adulteration testing |
Storage Requirements | 2° to 10°C |
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity & Availability | ||||
23-424-4538
|
Sciteck
103 |
Aldehyde Positive Control | 60mL |
Each for $344.00
|
|
23-424-4539
|
Sciteck
104 |
Aldehyde Negative Control | 60mL |
Each for $344.00
|
|
23-424-4537
|
Sciteck
123 |
Aldehyde Low Calibrator | 60mL |
Each for $344.00
|
|
Description
- For in vitro diagnostic use to detect presence of aldehydes in solution
- Analysis based on an indicator reaction which gives a change of absorbance in presence of UrinAid, and/or Aldehydes (Note: UrinAid is a commercial adulterant and is not a by-product of human metabolism)
- May be used on any freshly voided urine specimen or urine collected under special circumstances (e.g. first-morning specimens, post-prandial urine)
Sciteck Specimen Validity Testing (SVT)
- In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
- Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
- Adulteration testing products to include extensive SVT adulteration and EIA Drugs of Abuse single and multi-assay(s) (4-in-1) reagent line
- SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
- AdultaCheck for on-site adulteration products
Specifications
Human Urine | |
In-vitro diagnostic use to detect the presence of Aldehydes in solution, forensic/toxicology use | |
Automated system for drugs of abuse adulteration testing | |
2° to 10°C |
U.S. Patent No. 5,464,775. Other U.S. and Foreign Patents Pending
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title