Learn More
Sciteck™ SVT™ Adulteration Chromate Reagent
For use with automated systems for drugs of abuse adulteration testing
Manufacturer:  Sciteck™ 60002
 View more versions of this product
Catalog No. 23-424-4554
Add to cart
Description
Description
  • For in vitro diagnostic use to detect presence of chromates in solution
  • Analysis based on an indicator reaction which gives a change of absorbance in presence of chromates
  • May be used on any freshly voided urine specimen or urine collected under special circumstances (e.g. first-morning specimens, post-prandial urine)
  • Use with Zero Calibrator (23-424-4545)
Sciteck™ Specimen Validity Testing (SVT™)
  • In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
  • Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
  • Adulteration testing products to include extensive SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line
  • SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
  • AdultaCheck™ for on-site adulteration products
Specifications
Specifications
Human Urine
2° to 10°C
In-vitro diagnostic use to quantitatively determine the presence of chromates in Urine, forensic/toxicology use
Automated system for drugs of abuse adulteration testing
60mL
Chromate Calibrator, 100mg/dL
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
U.S. and Foreign Patents and Patents Pending