Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sciteck™ SVT™ Adulteration Chromate Reagent

For use with automated systems for drugs of abuse adulteration testing

$442.20 - $929.00

Specifications

Source Human Urine
Intended Use In-vitro diagnostic use to quantitatively determine the presence of chromates in Urine, forensic/toxicology use
For Use With (Equipment) Automated system for drugs of abuse adulteration testing
Storage Requirements 2° to 10°C
View More Specs

Products 5
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
23-424-4554
View Documents
Sciteck™
60002
Chromate Calibrator, 100mg/dL 60mL
Each for $442.20
Add to cart
 
23-424-4552
View Documents
Sciteck™
60003
Chromate Calibrator, 5mg/dL 60mL
Each for $442.20
Add to cart
 
23-424-4553
View Documents
Sciteck™
60001
Chromate Calibrator, 10mg/dL 60mL
Each for $442.20
Add to cart
 
23-424-4555
View Documents
Sciteck™
60004
Chromate Calibrator, 50mg/dL 60mL
Each for $442.20
Add to cart
 
23-424-4551
View Documents
Sciteck™
600
Chromate Reagent R(1) 750mL
Each for $929.00
Add to cart
 
Description

Description

  • For in vitro diagnostic use to detect presence of chromates in solution
  • Analysis based on an indicator reaction which gives a change of absorbance in presence of chromates
  • May be used on any freshly voided urine specimen or urine collected under special circumstances (e.g. first-morning specimens, post-prandial urine)
  • Use with Zero Calibrator (23-424-4545)

Sciteck™ Specimen Validity Testing (SVT™)

  • In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
  • Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
  • Adulteration testing products to include extensive SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line
  • SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
  • AdultaCheck™ for on-site adulteration products

Specifications

Specifications

Human Urine
In-vitro diagnostic use to quantitatively determine the presence of chromates in Urine, forensic/toxicology use
Automated system for drugs of abuse adulteration testing
2° to 10°C
SDS

U.S. and Foreign Patents and Patents Pending

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit