Learn More
Sciteck™ SVT™ Adulteration Creatinine Calibrators
For use with automated systems for drugs of abuse adulteration testing
Manufacturer:  SCITECK DIAGNOSTICS INC 13201
 View more versions of this product
Catalog No. 23-424-4534
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Creatinine calibrator
20mg/dL
In-vitro diagnostic use in the quantitative determination of creatinine in Serum and Urine
2° to 10°C
Serum and Urine
505nm
Automated system for drugs of abuse adulteration testing
60mL