Sciteck™ SVT™ Adulteration Creatinine Calibrators

For use with automated systems for drugs of abuse adulteration testing

$71.03 - $73.87

Specifications

For Use With Equipment Automated system for drugs of abuse adulteration testing
For Use With Application In-vitro diagnostic use in the quantitative determination of creatinine in Serum and Urine
Wavelength 505nm
Source Serum and Urine
View More Specs
  • Products 3
  • Description & Specifications
Products
Catalog Number Mfr. No. Product Type Concentration Quantity Price Quantity & Availability    

23-424-4534

Sciteck Diagnostics
132-01
Creatinine calibrator 20mg/dL 60mL Each for $73.87
Check Availability
Add to Cart

23-424-4603

Sciteck Diagnostics
136-01
Creatinine calibrator 100mg/dL 60mL Each for $71.03
Check Availability
Add to Cart

23-424-4535

Sciteck Diagnostics
137-01
Creatinine calibrator 400mg/dL 60mL Each for $71.81
Check Availability
Add to Cart
Descriptions & Specifications

Related Products