Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sciteck™ SVT™ Adulteration Halogen (Bleach) Reagent
For laboratory use only to determine halogen (e.g. bleach, etc.) content of a solution
Manufacturer: Sciteck™ 41003
Description
- If present, reacts with reagent containing a substituted biphenyl compound to yield a color complex
- Read results on an automated analyzer, or manually, at 660nm
- May be used on freshly voided urine specimen or urine collected under special conditions (e.g. first-morning specimens and post-prandial urine)
- Use with Zero Calibrator (23-424-4545)
Sciteck™ Specimen Validity Testing (SVT™)
- In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
- Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
- Adulteration testing products to include extensive SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line
- SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
- AdultaCheck™ for on-site adulteration products
Specifications
Automated analyzer system | |
60mL | |
2° to 10°C | |
660nm |
Laboratory use to determine Halogen (e.g. bleach, etc.) content of a solution | |
Human Urine | |
Halogen High Control |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
U.S. and Foreign Patents and Patents Pending: 6.537,823.