Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sciteck™ SVT™ Adulteration Halogen (Bleach) Reagent

For laboratory use only to determine halogen (e.g. bleach, etc.) content of a solution

$201.00 - $1114.00

Specifications

Source Human Urine
Storage Requirements 2° to 10°C
Wavelength 660nm
View More Specs

Products 5
Catalog Number Mfr. No. Type Intended Use For Use With (Equipment) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Type Intended Use For Use With (Equipment) Quantity Price Quantity & Availability  
23-424-4546
View Documents
Sciteck™
400
Halogen (Bleach) Reagent R1 Semi quantitative detection of Halogen(s) in Urine submitted for drugs of abuse testing, forensic/toxicology use Automated analyzer system 900mL
Each for $1,114.00
Add to cart
 
23-424-4547
View Documents
Sciteck™
40001
Halogen Calibrator Semi quantitative detection of Halogen(s) in Urine submitted for drugs of abuse testing, forensic/toxicology use Refractometer (specific gravity), pH meter (pH), automated analyzer 60mL
Each for $441.10
Add to cart
 
23-424-4548
View Documents
Sciteck™
40002
Halogen Positive Control Semi quantitative detection of Halogen(s) in Urine submitted for drugs of abuse testing, forensic/toxicology use Refractometer (specific gravity), pH meter (pH), automated analyzer 60mL
Each for $201.00
Add to cart
 
23-424-4550
View Documents
Sciteck™
41002
Halogen Low Control Semi quantitative detection of Halogen(s) in Urine submitted for drugs of abuse testing, forensic/toxicology use Refractometer (specific gravity), pH meter (pH), automated analyzer 60mL
Each for $201.00
Add to cart
 
23-424-4549
View Documents
Sciteck™
41003
Halogen High Control Laboratory use to determine Halogen (e.g. bleach, etc.) content of a solution Automated analyzer system 60mL
Each for $201.00
Add to cart
 
Description

Description

  • If present, reacts with reagent containing a substituted biphenyl compound to yield a color complex
  • Read results on an automated analyzer, or manually, at 660nm
  • May be used on freshly voided urine specimen or urine collected under special conditions (e.g. first-morning specimens and post-prandial urine)
  • Use with Zero Calibrator (23-424-4545)

Sciteck™ Specimen Validity Testing (SVT™)

  • In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
  • Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
  • Adulteration testing products to include extensive SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line
  • SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
  • AdultaCheck™ for on-site adulteration products

Specifications

Specifications

Human Urine
2° to 10°C
660nm
SDS

U.S. and Foreign Patents and Patents Pending: 6.537,823.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit