Sciteck™ SVT™ Adulteration Halogen (Bleach) Reagent

For laboratory use only to determine halogen (e.g. bleach, etc.) content of a solution

Manufacturer: SCITECK DIAGNOSTICS INC 41002

 View more versions of this product

Catalog No. 23-424-4550

  Check Availability
Add to Cart

Disclaimers:

U.S. and Foreign Patents and Patents Pending: 6.537,823.


    Specifications

    For Use With (Application) Semi quantitative detection of Halogen(s) in Urine submitted for drugs of abuse testing, forensic/toxicology use
    Product Type Halogen low control
    Wavelength 660nm
    For Use With (Equipment) Refractometer (specific gravity), pH meter (pH), automated analyzer
    Certifications/Compliance CAP FDT the Gold Standard, SAMHSA, HHS CLIA
    Storage Requirements 2° to 10°C
    Quantity 60mL,60mL
    Source Human Urine

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit