Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sciteck™ SVT™ Adulteration Nitrite Calibrators

For laboratory use only to determine nitrite content of a solution

$446.60

Specifications

Source Human Urine
Intended Use In-vitro diagnostic use to quantitatively determine the nitrite content of a solution, forensic/toxicology use
Storage Requirements 2° to 10°C
View More Specs

Products 4
Catalog Number Mfr. No. Concentration Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Type Quantity Price Quantity & Availability  
23-424-4541
View Documents
Sciteck™
211AD06
200mg/dL Nitrite Calibrator 60mL
Each for $446.60
Add to cart
 
23-424-4543
View Documents
Sciteck™
211AD07
1000mg/dL Nitrite Calibrator 60mL
Each for $446.60
Add to cart
 
23-424-4544
View Documents
Sciteck™
211AD08
10000mg/dL Nitrite Calibrator 60mL
Each for $446.60
Add to cart
 
23-424-4542
View Documents
Sciteck™
211AD09
500mg/dL Nitrite Calibrator 60mL
Each for $446.60
Add to cart
 
Description

Description

  • Non-corrosive
  • If present, reacts with reagent aromatic armine to form a diazonium salt, which couples with an indicator to yield a color complex
  • May be used on freshly voided urine specimen or urine collected under special conditions (e.g. first-morning specimens and post-prandial urine)
  • Use with Zero Calibrator (23-424-4545)

Sciteck™ Specimen Validity Testing (SVT™)

  • In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
  • Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
  • Adulteration testing products to include extensive SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line
  • SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
  • AdultaCheck™ for on-site adulteration products

Specifications

Specifications

Human Urine
In-vitro diagnostic use to quantitatively determine the nitrite content of a solution, forensic/toxicology use
2° to 10°C
SDS

U.S. and Foreign Patents and Patents Pending: 5,759,860.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit