Sciteck™ SVT™ Adulteration Nitrite Calibrators

For laboratory use only to determine nitrite content of a solution

Manufacturer: SCITECK DIAGNOSTICS INC 211AD09

 View more versions of this product

Catalog No. 23-424-4542

  N/A
Add to Cart

Disclaimers:

U.S. and Foreign Patents and Patents Pending: 5,759,860.

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Product Type Nitrite calibrator
    Concentration 500mg/dL
    For Use With (Application) In-vitro diagnostic use to quantitatively determine the nitrite content of a solution, forensic/toxicology use
    For Use With (Equipment) Refractometer (specific gravity), pH meter (pH)
    Certifications/Compliance CAP FDT the Gold Standard, SAMHSA, HHS CLIA
    Storage Requirements 2° to 10°C
    Quantity 60mL,60mL
    Source Human Urine

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit