Sciteck™ SVT™ Adulteration Oxidant Reagent

Detects adulteration in urine by quantitative determination of oxidants in urine


 View all options for this product

Catalog No. 23-424-4558


This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Product Type Oxidant reagent R1
    Source Human Urine
    Wavelength 660nm
    For Use With (Application) Detects adulteration in Urine by the quantitative determination of Oxidants, forensic/toxicology use
    Storage Requirements 2° to 10°C
    Quantity 750mL

    • Non-corrosive
    • Not for use in diagnosing disease or illness to effect treatment, and/or cure
    • For forensic/toxicology use only
    • Oxidants include nitrite, chromate, bleach, etc.
    • Colorimetric: endpoint
    • If present, reacts with oxidant reagent containing a substituted benzene compound and forms a color complex
    • Results readable at 660nm
    • May be used on freshly voided urine specimen or urine collected under special conditions (e.g. first-morning specimens and post-prandial urine)
    • Use with Nitrite Calibrators (23-424-4541, -4542); Zero Calibrator (23-424-4545)

    Sciteck™ Specimen Validity Testing (SVT™)

    • In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
    • Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
    • Adulteration testing products to include extensive SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line
    • SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
    • AdultaCheck™ for on-site adulteration products

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit