Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sciteck™ SVT™ Adulteration Peroxidase Reagent

For in vitro diagnostic use to determine peroxidase in urine

$1114.00

Specifications

Source Human Urine
Intended Use In-vitro diagnostic use to semi quantitatively determine the presence of Peroxidase in Urine
Storage Requirements 2° to 10°C
Wavelength 600nm
View More Specs

Products 1
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
23-424-4556
View Documents
Sciteck™
800
Peroxidase Reagent R1 and R2 750 and 150mL
Pack of 2 for $1,114.00
Add to cart
 
Description

Description

  • Colorimetric: endpoint
  • If present, reacts with peroxidase reagent containing a substituted benzene compound, activator and forms a color complex
  • Results readable at 600nm
  • May be used on freshly voided urine specimen or urine collected under special conditions (e.g. first-morning specimens and post-prandial urine)
  • Use with Zero Calibrator (23-424-4545)

Sciteck™ Specimen Validity Testing (SVT™)

  • In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
  • Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
  • Adulteration testing products to include extensive SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line
  • SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
  • AdultaCheck™ for on-site adulteration products

Specifications

Specifications

Human Urine
In-vitro diagnostic use to semi quantitatively determine the presence of Peroxidase in Urine
2° to 10°C
600nm
SDS

U.S. and Foreign Patents and Patents Pending.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit