Sciteck™ SVT™ Adulteration Peroxidase Reagent

For in vitro diagnostic use to determine peroxidase in urine

Manufacturer: SCITECK DIAGNOSTICS INC 800

 View more versions of this product

Catalog No. 23-424-4556

  Check Availability
Add to Cart

Disclaimers:

U.S. and Foreign Patents and Patents Pending.


    Specifications

    Product Type Peroxidase reagent R1 and R2
    Wavelength 600nm
    For Use With (Application) In-vitro diagnostic use to semi quantitatively determine the presence of Peroxidase in Urine
    Storage Requirements 2° to 10°C
    Quantity 750 and 150mL,750 and 150mL
    Source Human Urine

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit