Learn More
Sciteck™ SVT™ Adulteration pH Reagent
For in vitro diagnostic use to determine pH of a solution
Manufacturer:  SCITECK DIAGNOSTICS INC 109
 View more versions of this product
Catalog No. 23-424-4516
  Check Availability
Add to cart
Disclaimers:
U.S. and Foreign Patents and Patents Pending: 6,537,823; 5,759,860.
Specifications Description & Specifications
Specifications
pH calibrator
5, 8
2° to 10°C
Human Urine
In-vitro diagnostic use to quantitatively determine the pH of a solution
2 x 60mL