Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sciteck™ SVT™ Adulteration Quality Control Series

For use in validity tests for pH, creatinine, nitrite, and oxidants

$236.50 - $620.00

Specifications

Certifications/Compliance CAP FDT the Gold Standard, SAMHSA, HHS CLIA
Source Human Urine
Intended Use Use in validity tests for pH, Creatinine, Nitrite, and Oxidants, for forensic/toxicology use
For Use With (Equipment) Refractometer (specific gravity), pH meter (pH), automated clinical chemistry analyzer
Storage Requirements 2° to 10°C
View More Specs

Products 4
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
23-424-4564
View Documents
Sciteck™
A4300
Quality Control Series level 1 60mL Each for $236.50
Add to cart
 
23-424-4565
View Documents
Sciteck™
A4400
Quality Control Series level II 60mL Each for $236.50
Add to cart
 
23-424-4567
View Documents
Sciteck™
A4500
Quality Control Series Control set 3 x 60mL Pack of 3 for $620.00
Add to cart
 
23-424-4566
View Documents
Sciteck™
A4200
Quality Control Series level III 60mL Each for $236.50
Add to cart
 
Description

Description

As required by proposed SAMHSA guidelines in human urine on automated clinical chemistry analyzers

  • Not for use in diagnosing disease or illness to effect treatment, and/or cure
  • For forensic/toxicology use only
  • Ready-use-to liquid controls for use on autoanalyzers or dry chemistry methods
  • Sold individually or in sets and may be used with any reagent lot

Sciteck™ Specimen Validity Testing (SVT™)

  • In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
  • Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
  • Adulteration testing products to include extensive SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line
  • SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
  • AdultaCheck™ for on-site adulteration products

Specifications

Specifications

CAP FDT the Gold Standard, SAMHSA, HHS CLIA
Use in validity tests for pH, Creatinine, Nitrite, and Oxidants, for forensic/toxicology use
2° to 10°C
Human Urine
Refractometer (specific gravity), pH meter (pH), automated clinical chemistry analyzer
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit