Learn More
Sciteck™ SVT™ Adulteration Quality Control Series
For use in validity tests for pH, creatinine, nitrite, and oxidants
Manufacturer:  SCITECK DIAGNOSTICS INC A4400
 View more versions of this product
Catalog No. 23-424-4565
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Quality control series level II
7
Refractometer (specific gravity), pH meter (pH), automated clinical chemistry analyzer
2° to 10°C
Human Urine
Use in validity tests for pH, Creatinine, Nitrite, and Oxidants, for forensic/toxicology use
CAP FDT the Gold Standard, SAMHSA, HHS CLIA
60mL