Sciteck™ SVT™ Adulteration Quality Control Series

For use in validity tests for pH, creatinine, nitrite, and oxidants

Manufacturer: SCITECK DIAGNOSTICS INC A4200

 View more versions of this product

Catalog No. 23-424-4566

  Check Availability
Add to Cart

    Specifications

    pH 10.5
    For Use With (Application) Use in validity tests for pH, Creatinine, Nitrite, and Oxidants, for forensic/toxicology use
    Quantity 60mL,60mL
    Product Type Quality control series level III
    For Use With (Equipment) Refractometer (specific gravity), pH meter (pH), automated clinical chemistry analyzer
    Certifications/Compliance CAP FDT the Gold Standard, SAMHSA, HHS CLIA
    Storage Requirements 2° to 10°C
    Source Human Urine

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit