Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sciteck™ SVT™ Adulteration SAM-Set™ Calibrator Series

For use in validity tests for pH, creatinine, nitrite, and refractometer (specific gravity)

$104.10 - $432.50

Specifications

Source Human Urine
Intended Use Forensic/toxicology use only
For Use With (Equipment) Refractometer (specific gravity), pH meter (pH), automated clinical chemistry analyzer
Storage Requirements 2° to 10°C
View More Specs

Products 6
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Type Quantity Price Quantity & Availability  
23-424-4568
View Documents
Sciteck™
A4101
SAM-Set™ Calibrator Series Buffers pH 3.0, 11.0 and 12.0 3 x 60mL
Pack of 3 for $276.50
Add to cart
 
23-424-4569
View Documents
Sciteck™
A4102
SAM-Set™ Calibrator Series Nitrite 350 and 565mg 2 x 60mL
Pack of 2 for $399.50
Add to cart
 
23-424-4575
View Documents
Sciteck™
A41040
SAM-Set™ Calibrator Series Refractometer 1.0000 60mL
Each for $104.10
Add to cart
 
23-424-4571
View Documents
Sciteck™
A4103S
SAM-Set™ Calibrator Series Creatinine 3, 5, 7, 12, 23 5 x 60mL
Pack of 5 for $432.50
Add to cart
 
23-424-4576
View Documents
Sciteck™
A41042
SVT™ Specific Gravity Calibrator - Refractometer 1.0200 60mL
Each for $104.10
Add to cart
 
23-424-4574
View Documents
Sciteck™
A41043
SVT™ Specific Gravity Calibrator - Refractometer 1.0030 60mL
Each for $104.10
Add to cart
 
Description

Description

As required in proposed SAMHSA guidelines in human urine on automated clinical chemistry analyzers

  • Not for use in diagnosing disease or illness to effect treatment, and/or cure
  • For forensic/toxicology use only
  • Sold in sets per consituent and may be used with any reagent lot

Sciteck™ Specimen Validity Testing (SVT™)

  • In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
  • Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
  • Adulteration testing products to include extensive SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line
  • SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
  • AdultaCheck™ for on-site adulteration products

Specifications

Specifications

Human Urine
Forensic/toxicology use only
Refractometer (specific gravity), pH meter (pH), automated clinical chemistry analyzer
2° to 10°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit