Sciteck™ SVT™ Adulteration SAM-Set™ Calibrator Series

For use in validity tests for pH, creatinine, nitrite, and refractometer (specific gravity)

Manufacturer: SCITECK DIAGNOSTICS INC A41035

 View more versions of this product

Catalog No. 23-424-4570

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type SAM-Set™ calibrator series Creatinine 5.0mg/dL
    Source Human Urine
    For Use With (Application) Forensic/toxicology use only
    For Use With (Equipment) Refractometer (specific gravity), pH meter (pH), automated clinical chemistry analyzer
    Certifications/Compliance CAP FDT the Gold Standard, SAMHSA, HHS CLIA
    Storage Requirements 2° to 10°C
    Quantity 60mL

    As required in proposed SAMHSA guidelines in human urine on automated clinical chemistry analyzers

    • Not for use in diagnosing disease or illness to effect treatment, and/or cure
    • For forensic/toxicology use only
    • Sold in sets per consituent and may be used with any reagent lot

    Sciteck™ Specimen Validity Testing (SVT™)

    • In accordance with SAMHSA's FWDTP guidelines as specified by Executive Order 12564 and delineated in the Federal Register Vol. 53 FR 11970-11989 and Vol. 59 FR 29908-29931; and other federally mandated programs (DOT, DOE, NRC and FHWA) for employment drug screening
    • Sciteck evolved from Chimera Research, providing liquid reagents to laboratories for specimen validity testing in urine specimens submitted for DAU testing since 1989
    • Adulteration testing products to include extensive SVT adulteration and EIA™ Drugs of Abuse single and multi-assay(s) (4-in-1™) reagent line
    • SVT line consists of assays for pH, specific gravity, nitrite (non-corrosive), aldehydes, oxidants, chromate, creatinine, and halogens (bleach)
    • AdultaCheck™ for on-site adulteration products

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit