Sciteck™ SVT™ Adulteration SAM-Set™ Calibrator Series

For use in validity tests for pH, creatinine, nitrite, and refractometer (specific gravity)

Manufacturer: SCITECK DIAGNOSTICS INC A4103S

 View more versions of this product

Catalog No. 23-424-4571

  Check Availability
Add to Cart

    Specifications

    Product Type SAM-Set™ calibrator series Creatinine 3, 5, 7, 12, 23
    For Use With (Application) Forensic/toxicology use only
    For Use With (Equipment) Refractometer (specific gravity), pH meter (pH), automated clinical chemistry analyzer
    Certifications/Compliance CAP FDT the Gold Standard, SAMHSA, HHS CLIA
    Storage Requirements 2° to 10°C
    Quantity 5 x 60mL,5 x 60mL
    Source Human Urine

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit